1-Boc-4-(hydroxymethyl)piperidine 98%

BLDPBD13499-100GEA 47 EUR
BLDPBD13499-0.025G BLDPBD13499-100G BLDPBD13499-25G BLDPBD13499-500G BLDPBD13499-5G BLDPBD13499-10G BLDPBD13499-0.5G
1-Boc-4-(hydroxymethyl)piperidine 98%
1-Boc-4-(hydroxymethyl)piperidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₁H₂₁NO₃
MW: 215,29 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02094488
Núm. CAS: 123855-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR