3,5-Diiodo-4-methoxybenzonitrile 97%

BLDPBD134828-1GEA 116 EUR
BLDPBD134828-1G BLDPBD134828-250MG
3,5-Diiodo-4-methoxybenzonitrile 97%
3,5-Diiodo-4-methoxybenzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₅I₂NO
MW: 384,9400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00144497
Núm. CAS: 3336-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR