(2-Carboxyethyl)triphenylphosphonium chloride 98+%
|
|
BLDPBD1345-100GEA
195
EUR
BLDPBD1345-0.025G
BLDPBD1345-100G
BLDPBD1345-25G
BLDPBD1345-5G
(2-Carboxyethyl)triphenylphosphonium chloride 98+%
(2-Carboxyethyl)triphenylphosphonium chloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₂₁H₂₀ClO₂P MW: 370,81 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00185663 Núm. CAS: 36626-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...