Ethyl 2-(6-(ethyl(2-hydroxypropyl)amino)pyridazin-3-yl)hydrazine-1-carboxylate 95%

BLDPBD134097-100MGEA 160 EUR
BLDPBD134097-100MG BLDPBD134097-250MG
Ethyl 2-(6-(ethyl(2-hydroxypropyl)amino)pyridazin-3-yl)hydrazine-1-carboxylate 95%
Ethyl 2-(6-(ethyl(2-hydroxypropyl)amino)pyridazin-3-yl)hydrazine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₂₁N₅O₃
MW: 283,3268 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00865833
Núm. CAS: 64241-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR