(4E,8E)-(E)-3,7-Dimethylocta-2,6-dien-1-yl 5,9,13-trimethyltetradeca-4,8,12-trienoate 95%

BLDPBD131965-100MGEA 21 EUR
BLDPBD131965-100MG BLDPBD131965-250MG
(4E,8E)-(E)-3,7-Dimethylocta-2,6-dien-1-yl 5,9,13-trimethyltetradeca-4,8,12-trienoate 95%
(4E,8E)-(E)-3,7-Dimethylocta-2,6-dien-1-yl 5,9,13-trimethyltetradeca-4,8,12-trienoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₇H₄₄O₂
MW: 400,63 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00129086
Núm. CAS: 51-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR