2,3,3-Trimethyl-5-nitro-3H-indole 98%

BLDPBD13085-100MGEA 7 EUR
BLDPBD13085-100MG BLDPBD13085-1G BLDPBD13085-25G BLDPBD13085-250MG BLDPBD13085-5G BLDPBD13085-10G
2,3,3-Trimethyl-5-nitro-3H-indole 98%
2,3,3-Trimethyl-5-nitro-3H-indole

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₁₂N₂O₂
MW: 204,2252 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00447544
Núm. CAS: 3484-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR