(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-2-yl)propanoic acid 95%

BLDPBD130767-1GEA 412 EUR
BLDPBD130767-1G BLDPBD130767-250MG BLDPBD130767-100MG
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-2-yl)propanoic acid 95%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-2-yl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₈H₂₃NO₄
MW: 437,4865 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03427991
Núm. CAS: 507472-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR