2-(2-Chlorophenyl)-5,7-dihydroxy-8-((3S,4R)-3-hydroxy-1-methylpiperidin-4-yl)-4H-chromen-4-one hydrochloride 98%

BLDPBD130403-10MGEA 43 EUR
BLDPBD130403-10MG BLDPBD130403-5MG BLDPBD130403-100MG BLDPBD130403-50MG
2-(2-Chlorophenyl)-5,7-dihydroxy-8-((3S,4R)-3-hydroxy-1-methylpiperidin-4-yl)-4H-chromen-4-one hydrochloride 98%
2-(2-Chlorophenyl)-5,7-dihydroxy-8-((3S,4R)-3-hydroxy-1-methylpiperidin-4-yl)-4H-chromen-4-one hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₁H₂₁Cl₂NO₅
MW: 438,30 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD26792554
Núm. CAS: 131740-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR