Propyl heptanoate 98%

BLDPBD130094-100GEA 126 EUR
BLDPBD130094-100G BLDPBD130094-500G BLDPBD130094-25G BLDPBD130094-5G
Propyl heptanoate 98%
Propyl heptanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₂₀O₂
MW: 172,2646 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00048897
Núm. CAS: 7778-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR