2-(2-Hydroxyethoxy)ethyl 2-((3-(trifluoromethyl)phenyl)amino)benzoate 98%

BLDPBD129861-250MGEA 243 EUR
BLDPBD129861-250MG BLDPBD129861-100MG BLDPBD129861-50MG
2-(2-Hydroxyethoxy)ethyl 2-((3-(trifluoromethyl)phenyl)amino)benzoate 98%
2-(2-Hydroxyethoxy)ethyl 2-((3-(trifluoromethyl)phenyl)amino)benzoate
Formula: C₁₈H₁₈F₃NO₄
MW: 369,34 g/mol
Número MDL: MFCD00242806
Núm. CAS: 30544-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR