Ethyl 5-amino-4-phenylisoxazole-3-carboxylate 97%

BLDPBD129053-1GEA 110 EUR
BLDPBD129053-1G BLDPBD129053-250MG
Ethyl 5-amino-4-phenylisoxazole-3-carboxylate 97%
Ethyl 5-amino-4-phenylisoxazole-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₂H₁₂N₂O₃
MW: 232,23 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD06199347
Núm. CAS: 53983-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR