Ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate 95%

BLDPBD128431-250MGEA 45 EUR
BLDPBD128431-250MG BLDPBD128431-1G BLDPBD128431-100MG
Ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate 95%
Ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₁₀Cl₂O₃
MW: 261,1013 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD08458321
Núm. CAS: 72835-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR