Ethyl 2-propylpentanoate 95%

BLDPBD128415-1GEA 58 EUR
BLDPBD128415-1G BLDPBD128415-250MG BLDPBD128415-5G
Ethyl 2-propylpentanoate 95%
Ethyl 2-propylpentanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₂₀O₂
MW: 172,26 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00870991
Núm. CAS: 17022-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR