7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona 98%

BLDPBD12786-100GEA 225 EUR
BLDPBD12786-100G BLDPBD12786-5G BLDPBD12786-1G BLDPBD12786-25G BLDPBD12786-500G BLDPBD12786-10G
7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona 98%
7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₉BrO
MW: 225,08 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02179287
Núm. CAS: 32281-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR