4-(1H-Pyrazol-1-yl)benzaldehyde 97%

BLDPBD12719-10GEA 59 EUR
BLDPBD12719-10G BLDPBD12719-100G BLDPBD12719-5G BLDPBD12719-250MG BLDPBD12719-1G BLDPBD12719-25G
4-(1H-Pyrazol-1-yl)benzaldehyde 97%
4-(1H-Pyrazol-1-yl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₈N₂O
MW: 172,1833 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02681938
Núm. CAS: 99662-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR