(R)-2-(2-((tert-Butoxycarbonyl)amino)-2-methylpropanamido)-3-(1H-indol-3-yl)propanoic acid 97%

BLDPBD127011-250MGEA 25 EUR
BLDPBD127011-250MG BLDPBD127011-5G BLDPBD127011-100MG BLDPBD127011-1G
(R)-2-(2-((tert-Butoxycarbonyl)amino)-2-methylpropanamido)-3-(1H-indol-3-yl)propanoic acid 97%
(R)-2-(2-((tert-Butoxycarbonyl)amino)-2-methylpropanamido)-3-(1H-indol-3-yl)propanoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₀H₂₇N₃O₅
MW: 389,45 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08704784
Núm. CAS: 159634-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR