1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium 98%

Sinónimos: NAD+

BLDPBD126917-10GEA 54 EUR
BLDPBD126917-10G BLDPBD126917-100G BLDPBD126917-25G BLDPBD126917-1G BLDPBD126917-5G BLDPBD126917-250MG
1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium 98%
1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663,425 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00036253
Núm. CAS: 53-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR