4-Chloro-7-methoxyquinoline 98%

BLDPBD12686-10GEA 207 EUR
BLDPBD12686-10G BLDPBD12686-1G BLDPBD12686-25G BLDPBD12686-5G BLDPBD12686-100G BLDPBD12686-250MG
4-Chloro-7-methoxyquinoline 98%
4-Chloro-7-methoxyquinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₈ClNO
MW: 193,6300 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD06659036
Núm. CAS: 68500-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR