Dl-2-amino-1-hexanol 95%

BLDPBD126285-250MGEA 235 EUR
BLDPBD126285-250MG BLDPBD126285-1G BLDPBD126285-100MG
Dl-2-amino-1-hexanol 95%
Dl-2-amino-1-hexanol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₅NO
MW: 117,1894 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00009810
Núm. CAS: 5665-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR