Ethoxy(methyl)diphenylsilane 95%

BLDPBD125907-10GEA 306 EUR
BLDPBD125907-10G BLDPBD125907-5G BLDPBD125907-250MG BLDPBD125907-1G
Ethoxy(methyl)diphenylsilane 95%
Ethoxy(methyl)diphenylsilane

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₅H₁₈OSi
MW: 242,39 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00026752
Núm. CAS: 1825-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR