Diphenyl phosphate 97%
|
|
BLDPBD125832-500GEA
88
EUR
BLDPBD125832-500G
BLDPBD125832-5G
BLDPBD125832-1000G
BLDPBD125832-25G
BLDPBD125832-100G
Diphenyl phosphate 97%
Diphenyl phosphate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₂H₁₁O₄P MW: 250,19 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00003033 Núm. CAS: 838-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...