2-(Dimethylamino)acetohydrazide 98%

BLDPBD125544-5GEA 277 EUR
BLDPBD125544-5G BLDPBD125544-250MG BLDPBD125544-1G
2-(Dimethylamino)acetohydrazide 98%
2-(Dimethylamino)acetohydrazide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₄H₁₁N₃O
MW: 117,14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00272257
Núm. CAS: 55-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR