Dimethyl-trans-cyclohexane-1,4-dicarboxylate 96%

Proveedor: BLD PHARMATECH GMBH

BLDPBD125488-100GEA 90 EUR
BLDPBD125488-100G BLDPBD125488-25G BLDPBD125488-1G BLDPBD125488-500G BLDPBD125488-5G BLDPBD125488-10G
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 96%
Dimethyl-trans-cyclohexane-1,4-dicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₆O₄
MW: 200,23 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00063917
Núm. CAS: 3399-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR