Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate 98%

BLDPBD124389-100GEA 29 EUR
BLDPBD124389-100G BLDPBD124389-1000G BLDPBD124389-25G BLDPBD124389-5G BLDPBD124389-500G
Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate 98%
Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₈H₂₂Cl₂N₂O₂
MW: 369,2855 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01862249
Núm. CAS: 78213-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR