4-Cyclohexylbutan-1-ol 95%

BLDPBD12421-100MGEA 26 EUR
BLDPBD12421-100MG BLDPBD12421-1G BLDPBD12421-5G BLDPBD12421-250MG
4-Cyclohexylbutan-1-ol 95%
4-Cyclohexylbutan-1-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₂₀O
MW: 156,26 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001531
Núm. CAS: 4441-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR