Decyloleate 95%

BLDPBD123650-1GEA 915 EUR
BLDPBD123650-1G BLDPBD123650-100MG BLDPBD123650-250MG
Decyloleate 95%
Decyloleate

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₈H₅₄O₂
MW: 422,72 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00048401
Núm. CAS: 3687-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR