Diethyl (3-chloropropyl)malonate 95+%
|
|
BLDPBD12344-5GEA
45
EUR
BLDPBD12344-5G
BLDPBD12344-25G
BLDPBD12344-1G
Diethyl (3-chloropropyl)malonate 95+%
Diethyl (3-chloropropyl)malonate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₁₇ClO₄ MW: 236,70 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00009169 Núm. CAS: 18719-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...