(R)-6-Oxopiperidine-2-carboxylic acid 98%

BLDPBD123330-100MGEA 41 EUR
BLDPBD123330-100MG BLDPBD123330-1G BLDPBD123330-250MG
(R)-6-Oxopiperidine-2-carboxylic acid 98%
(R)-6-Oxopiperidine-2-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₉NO₃
MW: 143,1400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09025314
Núm. CAS: 72002-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR