1-Cyclopentenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD122948-5GEA 80 EUR
BLDPBD122948-5G BLDPBD122948-1G BLDPBD122948-10G BLDPBD122948-250MG BLDPBD122948-25G
1-Cyclopentenylboronic acid 98%
1-Cyclopentenylboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₅H₉BO₂
MW: 111,94 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD02179496
Núm. CAS: 850036-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR