Cyclopentadienyltitanium trichloride 98%

BLDPBD122912-250MGEA 26 EUR
BLDPBD122912-250MG BLDPBD122912-5G BLDPBD122912-25G BLDPBD122912-1G
Cyclopentadienyltitanium trichloride 98%
Cyclopentadienyltitanium trichloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅Cl₃Ti
MW: 214,28 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00003724
Núm. CAS: 1270-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR