3,7-Dimethyloct-6-en-1-yl formate 95+%

BLDPBD121835-100GEA 27 EUR
BLDPBD121835-100G BLDPBD121835-25G BLDPBD121835-0.5G BLDPBD121835-500G
3,7-Dimethyloct-6-en-1-yl formate 95+%
3,7-Dimethyloct-6-en-1-yl formate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₂₀O₂
MW: 184,2753 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00021050
Núm. CAS: 105-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR