((Chloromethyl)sulphinyl)benzene 95+%

BLDPBD120868-250MGEA 103 EUR
BLDPBD120868-250MG BLDPBD120868-1G BLDPBD120868-100MG
((Chloromethyl)sulphinyl)benzene 95+%
((Chloromethyl)sulphinyl)benzene

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₇H₇ClOS
MW: 174,64 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00042639
Núm. CAS: 7205-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR