Chloro(cyclopentadienyl)(triphenylphosphine)nickel(II) 98%

BLDPBD120715-1GEA 120 EUR
BLDPBD120715-1G BLDPBD120715-250MG BLDPBD120715-5G
Chloro(cyclopentadienyl)(triphenylphosphine)nickel(II) 98%
Chloro(cyclopentadienyl)(triphenylphosphine)nickel(II)

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₃H₁₇ClNiP
MW: 418,50 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02093655
Núm. CAS: 31904-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR