4-Methoxy-2-methylpyridine 98%

BLDPBD120577-5GEA 61 EUR
BLDPBD120577-5G BLDPBD120577-10G BLDPBD120577-1G BLDPBD120577-25G BLDPBD120577-100G
4-Methoxy-2-methylpyridine 98%
4-Methoxy-2-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉NO
MW: 123,1500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06253892
Núm. CAS: 24103-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR