4-Amino-N-(butylcarbamoyl)benzenesulphonamide 98%

BLDPBD120085-1GEA 269 EUR
BLDPBD120085-1G BLDPBD120085-250MG BLDPBD120085-100MG
4-Amino-N-(butylcarbamoyl)benzenesulphonamide 98%
4-Amino-N-(butylcarbamoyl)benzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₁H₁₇N₃O₃S
MW: 271,33 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00025344
Núm. CAS: 339-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR