EDTA calcium disodium salt 98+%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD119656-500GEA
31
EUR
BLDPBD119656-500G
BLDPBD119656-100G
BLDPBD119656-25G
EDTA calcium disodium salt 98+%
EDTA calcium disodium salt
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₁₂CaN₂Na₂O₈ MW: 374,2684 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00148919 Núm. CAS: 62-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...