Butyl cyclohexyl phthalate 95%

BLDPBD119139-1GEA 335 EUR
BLDPBD119139-1G BLDPBD119139-250MG BLDPBD119139-100MG
Butyl cyclohexyl phthalate 95%
Butyl cyclohexyl phthalate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₂₄O₄
MW: 304,3800 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00152312
Núm. CAS: 84-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR