5-Amino-1-methyl-3-phenyl-1H-pyrazole 98%
|
|
BLDPBD11904-1GEA
34
EUR
BLDPBD11904-0.025G
BLDPBD11904-1G
BLDPBD11904-5G
BLDPBD11904-25G
5-Amino-1-methyl-3-phenyl-1H-pyrazole 98%
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₀H₁₁N₃ MW: 173,22 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00067874 Núm. CAS: 10199-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...