(S)-3-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid 98%

BLDPBD117589-1GEA 119 EUR
BLDPBD117589-1G BLDPBD117589-250MG
(S)-3-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₈BrNO₄
MW: 344,20 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03427924
Núm. CAS: 500770-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR