Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate) 97%

BLDPBD117437-25GEA 9 EUR
BLDPBD117437-25G BLDPBD117437-100G
Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate) 97%
Tetraphenyl (propane-2,2-diylbis(4,1-phenylene)) bis(phosphate)

Storage condition: Sealed in dry, Room Temperature

Formula: C₃₉H₃₄O₈P₂
MW: 692,6300 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 5945-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR