Bis(4-methylbenzyl) oxalate 97%

BLDPBD117314-25GEA 16 EUR
BLDPBD117314-25G BLDPBD117314-500G BLDPBD117314-5G BLDPBD117314-0.025 BLDPBD117314-100G
Bis(4-methylbenzyl) oxalate 97%
Bis(4-methylbenzyl) oxalate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₁₈O₄
MW: 298,33 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00266809
Núm. CAS: 18241-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR