4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane 12.5-15.5% S

BLDPBD117252-1GEA 6 EUR
BLDPBD117252-1G BLDPBD117252-500G BLDPBD117252-10G BLDPBD117252-100G BLDPBD117252-25G BLDPBD117252-5G
4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane 12.5-15.5% S
4,4,13,13-Tetraethoxy-3,14-dioxa-8,9-dithia-4,13-disilahexadecane

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₈H₄₂O₆S₂Si₂
MW: 474,8235 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD05663903
Núm. CAS: 56706-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR