1,2-Bis(4-bromophenyl)disulphane 98%

BLDPBD116976-1GEA 72 EUR
BLDPBD116976-1G BLDPBD116976-250MG BLDPBD116976-25G BLDPBD116976-5G
1,2-Bis(4-bromophenyl)disulphane 98%
1,2-Bis(4-bromophenyl)disulphane

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₈Br₂S₂
MW: 376,12 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00017832
Núm. CAS: 5335-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR