Bis(2-methoxyethyl)phthalate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD116912-100GEA 40 EUR
BLDPBD116912-100G BLDPBD116912-500G BLDPBD116912-25G BLDPBD116912-1000G
Bis(2-methoxyethyl)phthalate 98%
Bis(2-methoxyethyl)phthalate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₈O₆
MW: 282,29 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00042842
Núm. CAS: 117-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR