Benzyloxyacetaldehyde dimethyl acetal 95%

BLDPBD116316-10GEA 54 EUR
BLDPBD116316-10G BLDPBD116316-25G BLDPBD116316-100G BLDPBD116316-5G
Benzyloxyacetaldehyde dimethyl acetal 95%
Benzyloxyacetaldehyde dimethyl acetal

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₆O₃
MW: 196,24 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00082233
Núm. CAS: 127657-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR