2-Bromo-1-(4-fluoro-3-methoxyphenyl)ethanone 97%

BLDPBD11602-250MGEA 23 EUR
BLDPBD11602-250MG
2-Bromo-1-(4-fluoro-3-methoxyphenyl)ethanone 97%
2-Bromo-1-(4-fluoro-3-methoxyphenyl)ethanone

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₈BrFO₂
MW: 247,0610 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03425169
Núm. CAS: 443914-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR