Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate 97%

BLDPBD115795-100MGEA 6 EUR
BLDPBD115795-100MG BLDPBD115795-250MG BLDPBD115795-1G BLDPBD115795-5G
Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate 97%
Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₂H₁₇N₃O₄
MW: 387,3881 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09029084
Núm. CAS: 139481-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR