(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one 98%

BLDPBD114723-50MGEA 343 EUR
BLDPBD114723-50MG BLDPBD114723-5MG BLDPBD114723-25MG
(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one 98%
(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one
Núm. CAS: 50906-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR