Methyl arachidate 98%
|
|
BLDPBD114599-1GEA
21
EUR
BLDPBD114599-1G
BLDPBD114599-25G
BLDPBD114599-100MG
BLDPBD114599-5G
BLDPBD114599-250MG
Methyl arachidate 98%
Methyl arachidate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₂₁H₄₂O₂ MW: 326,56 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00009014 Núm. CAS: 1120-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...