2-Amino-2-(2,6-difluorophenyl)acetic acid 98%

BLDPBD113885-250MGEA 38 EUR
BLDPBD113885-250MG BLDPBD113885-5G BLDPBD113885-1G BLDPBD113885-100MG
2-Amino-2-(2,6-difluorophenyl)acetic acid 98%
2-Amino-2-(2,6-difluorophenyl)acetic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₇F₂NO₂
MW: 187,14 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07786876
Núm. CAS: 244187-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR